Learn More
Hampton Research MEMBFAC NO48 185ML
SDP
Catalog No. 501799
Encompass
HR2-920-48/MembFac #48 (D12), 185 ml - 0.1 M Sodium citrate tribasic dihydrate, 0.1 M TRIS hydrochloride pH 8.5, 5% v/v Polyethylene glycol 400. Please note items from this supplier are non-returnable.
Catalog No. 50-179-9 Supplier Hampton Research Supplier No. HR292048
HR2-920-48/MembFac #48 (D12), 185 ml - 0.1 M Sodium citrate tribasic dihydrate, 0.1 M TRIS hydrochloride pH 8.5, 5% v/v Polyethylene glycol 400. Please note items from this supplier are non-returnable.