Learn More
Crescent Chemical Co Inc MEMBRA-CEL DIALYSIS TUBING
SDP
Supplier:  Crescent Chemical Co Inc SE44313.01
Encompass_Preferred
MEMBRA-CEL dialysis tubing renerated cellulose, diameter 16 mm Packed dry , made from regenerated cellulose (RC). With glycerol as protection for embrittlement, which can be easily removed by soaking in water. Contain low level of heavy metal and sulfide impurities. Highly r
Catalog No. 50-249-007