Learn More
Crescent Chemical Co Inc MEMBRA-CEL DIALYSIS TUBING
SDP
Supplier:  Crescent Chemical Co Inc SE44310.02
Encompass_Preferred
MEMBRA-CEL dialysis tubing RC, diameter 16 mm Packed dry , made from regenerated cellulose (RC). With glycerol as protection for embrittlement, which can be easily removed by soaking in water. Contain low level of heavy metal and sulfide impurities. Highly resistant against
Catalog No. 50-249-002