Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BPS Bioscience Inc Membrane-Bound TNFα CHO Cell Line, 2 vials, >1 x 10^6 cells per vial, adherent, Mycoplasma negative
SDP

Catalog No. NC3782737
Encompass

The Membrane-Bound TNFα CHO Cell Line is a clonal Chinese Hamster Ovary (CHO) cell line designed to express human membrane-bound Tumor Necrosis Factor alpha (mTNFα), pivotal for research in antibody-dependent cellular cytotoxicity (ADCC) and TNFα signaling pathways.

  • Cell type: CHO, epithelial-like, adherent
  • Format: 2 vials of frozen cells
  • Quantity per vial: >1 x 10^6 cells in 1 ml Cell Freezing Medium
  • Mycoplasma status: Negative
  • Application: Measure anti-TNFα antibody ADCC responses
  • Application: Measure antibody binding affinity to mTNFα
  • Transduction agent: mTNFα Lentivirus (#78955)
  • Sub-cultivation ratio: 1:6 to 1:8
  • Passage frequency: Once or twice a week

Catalog No. NC3782737 Supplier BPS Bioscience Inc Supplier No. 78971
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

The Membrane-Bound TNFα CHO Cell Line is a clonal Chinese Hamster Ovary (CHO) cell line designed to express human membrane-bound Tumor Necrosis Factor alpha (mTNFα), pivotal for research in antibody-dependent cellular cytotoxicity (ADCC) and TNFα signaling pathways.

  • Cell type: CHO, epithelial-like, adherent
  • Format: 2 vials of frozen cells
  • Quantity per vial: >1 x 10^6 cells in 1 ml Cell Freezing Medium
  • Mycoplasma status: Negative
  • Application: Measure anti-TNFα antibody ADCC responses
  • Application: Measure antibody binding affinity to mTNFα
  • Transduction agent: mTNFα Lentivirus (#78955)
  • Sub-cultivation ratio: 1:6 to 1:8
  • Passage frequency: Once or twice a week

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.