Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Double Elastic Membrane Box Rectangular, 250 x 200 mm, 3371 g Capacity
SDP

Catalog No. 5027118
Encompass_Preferred

Our unique membrane storage boxes for the transfer, storage and shipping of many delicate items. These plastic containers, with a double elastic membrane on the top and bottom, offer security for all of your sensitive items. Once your item is placed in the box the elastic membrane conforms around the enclosed object and protects it from any movement.
Dimensions: 250 x 200 mm
Height: 200 mm
Capacity: 3371 g
Use: 157 x 135 mm
Depth: 135 mm

Catalog No. 50-271-18 Supplier Electron Microscopy Sciences Supplier No. 6430445
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Our unique membrane storage boxes for the transfer, storage and shipping of many delicate items. These plastic containers, with a double elastic membrane on the top and bottom, offer security for all of your sensitive items. Once your item is placed in the box the elastic membrane conforms around the enclosed object and protects it from any movement.
Dimensions: 250 x 200 mm
Height: 200 mm
Capacity: 3371 g
Use: 157 x 135 mm
Depth: 135 mm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.