Learn More
Electron Microscopy Sciences Double Elastic Membrane Box Rectangular, 200 mm
SDP
Catalog No. 501926918
Encompass_Preferred
Our unique membrane storage boxes for the transfer, storage and shipping of many delicate items. These plastic containers, with a double elastic membrane on the top and bottom, offer security for all of your sensitive items. Once your item is placed in the box the elastic membrane conforms around the enclosed object and protects it from any movement.
Dimensions: 200 x 150 mm
Height: 50 mm
Catalog No. 50-192-6918 Supplier Electron Microscopy Sciences Supplier No. 6430459
Our unique membrane storage boxes for the transfer, storage and shipping of many delicate items. These plastic containers, with a double elastic membrane on the top and bottom, offer security for all of your sensitive items. Once your item is placed in the box the elastic membrane conforms around the enclosed object and protects it from any movement.
Dimensions: 200 x 150 mm
Height: 50 mm