Learn More
Electron Microscopy Sciences Double Elastic Membrane Box Square
SDP
Supplier:  Electron Microscopy Sciences 6430426
Encompass_Preferred
Our unique membrane storage boxes for the transfer, storage and shipping of many delicate items. These plastic containers, with a double elastic membrane on the top and bottom, offer security for all of your sensitive items. Once your item is placed in the box the elastic membrane conforms around the enclosed object and protects it from any movement.
Dimensions: 200 x 200 mm
Height: 50 mm
Catalog No. 50-192-6907