Learn More
LICORbio Membrane Carrier for the Odyssey Sa Imaging System
Catalog No. NC1956210
Encompass_Preferred
The glass membrane carrier for the Odyssey Sa Imaging System. This carrier is used for western blotting methods and other analysis requiring scans of membranes.
Catalog No. NC1956210 Supplier LICORbio Supplier No. 92673000
The glass membrane carrier for the Odyssey Sa Imaging System. This carrier is used for western blotting methods and other analysis requiring scans of membranes.