Learn More
Cusabio Technology LLC membrane cofactor protein MCP
Catalog No. 5000049764
Encompass_Preferred
This kit detects CD46 (Human membrane cofactor protein MCP) in serum plasma and tissue homogenates using a sandwich ELISA technique It offers a detection range of 3 12 ng/mL to 200 ng/mL with a sensitivity of 0 78 ng/mL enabling precise quantitative analysis The assay requires a sample volume of 50-100 L and involves an incubation period of 1-5 hours Measurements are performed at 450 nm absorbance ensuring reliable and reproducible results for research or diagnostic applications Designed for flexibility the kit accommodates multiple biological sample types while maintaining high specificity and accuracy in detecting CD46 levels
Catalog No. 50-000-49764 Supplier Cusabio Technology LLC Supplier No. CSBE09216H
This kit detects CD46 (Human membrane cofactor protein MCP) in serum plasma and tissue homogenates using a sandwich ELISA technique It offers a detection range of 3 12 ng/mL to 200 ng/mL with a sensitivity of 0 78 ng/mL enabling precise quantitative analysis The assay requires a sample volume of 50-100 L and involves an incubation period of 1-5 hours Measurements are performed at 450 nm absorbance ensuring reliable and reproducible results for research or diagnostic applications Designed for flexibility the kit accommodates multiple biological sample types while maintaining high specificity and accuracy in detecting CD46 levels