Learn More
Description
- For use with the Fisherbrand Pipet Controller
- 0.2 μm Hydrophobic PTFE
- Pack of 5 replacement Membrane Filters
Specifications
Specifications
| Description | Replacement parts only |
| Material | PTFE |
| Volume (Metric) | 0.20 μm |
| For Use With (Equipment) | Fisherbrand Pipet Controller (FB14955202) |
| Quantity | 5 Pieces |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.