Learn More
Molecular Devices Molecular Devices Filter Kit, Standard, 7 Filters, 2 Slides
Catalog No. 5000014445
Encompass_Preferred
Replacement standard filter kit for the Molecular Devices SpectraMax iD5 multi-mode microplate reader. The kit provides a set of optical filters and slides used to configure the reader for common filter-based detection modes.
  • Contains seven filters and two filter slides.
  • Provides configurable filters for common fluorescence and filter-based detection modes.
  • Supplied as a standard kit for instrument configuration and replacement of worn filters.
  • Includes components intended for installation as instrument accessory slides and filter holders.
Catalog No. 50-000-14445 Supplier Molecular Devices Supplier No. 65900152
Replacement standard filter kit for the Molecular Devices SpectraMax iD5 multi-mode microplate reader. The kit provides a set of optical filters and slides used to configure the reader for common filter-based detection modes.
  • Contains seven filters and two filter slides.
  • Provides configurable filters for common fluorescence and filter-based detection modes.
  • Supplied as a standard kit for instrument configuration and replacement of worn filters.
  • Includes components intended for installation as instrument accessory slides and filter holders.