Learn More
YSI 3P LifeScience L-Lactate Membrane Kit (4 each)
Catalog No. 50200113
Encompass
For in-vitro diagnostic use with YSI enzyme-based lactate analyzers (YSI 1500, 2300, 2700, 2900 series and 7100 analyzers) labeled for clinical use. Also for non-diagnostic use. Reactive ingredients: immobilized L-Lactate oxidase. Store membranes refrigerated; do not freeze. Return plastic holder with unused membranes to foil pouch. Discard when expiration date is past. Stability of enzyme preparation is variable. See appropriate YSI instruction manual for membrane activity tests. US patent 3979274. NOTE: Requires 2-day shipping during summer months.
Catalog No. 50-200-113 Supplier YSI 3P LifeScience Supplier No. 2329
For in-vitro diagnostic use with YSI enzyme-based lactate analyzers (YSI 1500, 2300, 2700, 2900 series and 7100 analyzers) labeled for clinical use. Also for non-diagnostic use. Reactive ingredients: immobilized L-Lactate oxidase. Store membranes refrigerated; do not freeze. Return plastic holder with unused membranes to foil pouch. Discard when expiration date is past. Stability of enzyme preparation is variable. See appropriate YSI instruction manual for membrane activity tests. US patent 3979274. NOTE: Requires 2-day shipping during summer months.