Learn More
Echelon Biosciences Research Labs MEMBRANE LIPID ARRAYS 5 pack
SDP
Supplier:  Echelon Biosciences Research Labs P60035PK
Encompass
Membrane Lipid Arrays are 4 x 5 cm hydrophobic membranes that have been spotted with a concentration gradient of eight different biologically important lipids found in cell membranes. They are used to determine the relative degree of binding of your protein of interest towards one or more of these lipids in a simple protein lipid overlay assay. Bound protein is then detected by an antibody against the protein or affinity tag. This item is non returnable
Catalog No. 50-720-659