Learn More
Echelon Biosciences Research Labs MEMBRANE LIPID STRIPS 2 pack
SDP
Catalog No. 50720657
Encompass
Membrane Lipid Strips are 2 x 6 cm hydrophobic membranes that have been spotted with 100 pmol of fifteen different biologically important lipids found in cell membranes. They are used to determine how your protein of interest interacts with one or more of these lipids in a simple protein-lipid overlay assay. Bound protein is then detected by an antibody against the protein or affinity tag. This item is non returnable
This item is not returnable. View return policy
Catalog No. 50-720-657 Supplier Echelon Biosciences Research Labs Supplier No. P60022PK
Membrane Lipid Strips are 2 x 6 cm hydrophobic membranes that have been spotted with 100 pmol of fifteen different biologically important lipids found in cell membranes. They are used to determine how your protein of interest interacts with one or more of these lipids in a simple protein-lipid overlay assay. Bound protein is then detected by an antibody against the protein or affinity tag. This item is non returnable