Learn More
Molecular Devices Molecular Devices Fatty Acid Uptake Assay, Homogeneous Fluorescence, 96/384-well
Catalog No. 5000014397
Encompass_Preferred
The QBT Fatty Acid Uptake Assay Kit is a one-step, homogeneous fluorescence assay for monitoring cellular uptake of fatty acids and screening fatty acid transport activity in live cells. It enables real-time kinetic measurements in 96- and 384-well plate formats and is optimized for high-throughput workflows.
  • One-step homogeneous assay format for streamlined workflow.
  • Uses a BODIPY-dodecanoic acid fluorescent fatty acid analog for physiological relevance.
  • Proprietary quench dye minimizes extracellular signal for improved signal-to-background.
  • Compatible with 96-well and 384-well plates for high-throughput screening.
  • Pre-optimized reagents supplied to reduce assay development time.
  • Eliminates need for radioactive tracers, reducing disposal and safety concerns.
Catalog No. 50-000-14397 Supplier Molecular Devices Supplier No. R8132
Add to Cart
Add to Cart
The QBT Fatty Acid Uptake Assay Kit is a one-step, homogeneous fluorescence assay for monitoring cellular uptake of fatty acids and screening fatty acid transport activity in live cells. It enables real-time kinetic measurements in 96- and 384-well plate formats and is optimized for high-throughput workflows.
  • One-step homogeneous assay format for streamlined workflow.
  • Uses a BODIPY-dodecanoic acid fluorescent fatty acid analog for physiological relevance.
  • Proprietary quench dye minimizes extracellular signal for improved signal-to-background.
  • Compatible with 96-well and 384-well plates for high-throughput screening.
  • Pre-optimized reagents supplied to reduce assay development time.
  • Eliminates need for radioactive tracers, reducing disposal and safety concerns.