Learn More
Biochain Institute Inc Membrane Protein - Alzheimer's Disease:Brain: Hippocampus, 0.1 mg/PK
SDP
Catalog No. 50180617
Encompass_Preferred
Tissue membrane protein is prepared from whole tissue homogenates and presents a consistent pattern on SDS-PAGE analysis. The protein is stored in a buffer with protease inhibitor cocktail.
Catalog No. 50-180-617 Supplier Biochain Institute Inc Supplier No. P3236052ALZ
Tissue membrane protein is prepared from whole tissue homogenates and presents a consistent pattern on SDS-PAGE analysis. The protein is stored in a buffer with protease inhibitor cocktail.