Learn More
Biochain Institute Inc Membrane Protein - Human Adult Normal Tissue: Brain: Cerebellum (right), 0.1 mg/PK
SDP
Catalog No. 50180529
Encompass_Preferred
Tissue membrane protein is prepared from whole tissue homogenates and presents a consistent pattern on SDS-PAGE analysis. The protein is stored in a buffer with protease inhibitor cocktail.
Catalog No. 50-180-529 Supplier Biochain Institute Inc Supplier No. P3234041
Tissue membrane protein is prepared from whole tissue homogenates and presents a consistent pattern on SDS-PAGE analysis. The protein is stored in a buffer with protease inhibitor cocktail.