Learn More
Biochain Institute Inc Membrane Protein - Human Adult Normal Tissue: Kidney, 0.1 mg/PK
SDP
Catalog No. 50180552
Encompass_Preferred
Tissue membrane protein is prepared from whole tissue homogenates and presents a consistent pattern on SDS-PAGE analysis. The protein is stored in a buffer with protease inhibitor cocktail.
Catalog No. 50-180-552 Supplier Biochain Institute Inc Supplier No. P3234142
Tissue membrane protein is prepared from whole tissue homogenates and presents a consistent pattern on SDS-PAGE analysis. The protein is stored in a buffer with protease inhibitor cocktail.