Learn More
Enzo Life Sciences Membrin monoclonal antibody (4HAD6) (200µg)
SDP
Supplier:  Enzo Life Sciences ADIVAMPT046F
Encompass_Preferred
Clone: 4HAD6. Host: Mouse. Isotype: IgG1. Immunogen: Recombinant rat Membrin protein with a His tag. Source: Purified from ascites. Species reactivity: Human, Rat, Rabbit, Porcine, Monkey, Hamster, Dog, Chicken. Applications: ICC, IHC (PS), WB. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 0.1MM PMSF and 50% glycerol.
Catalog No. 50-200-7689