Learn More
Enzo Life Sciences Membrin polyclonal antibody (25µg)
SDP
Supplier:  Enzo Life Sciences ADIVAPPT049C
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to a portion of rat membrin. UniProt ID: O35165. Source: Purified from rabbit serum. Species reactivity: Human, Mouse, Rat, Sheep, Rabbit, Porcine, Monkey, Hamster, Drosophila, Dog, Chicken, Bovine. Applications: IHC (PS), WB. Recommended Dilutions: Western Blot (2µg/ml, ECL) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Peptide immunoaffinity purified. Formulation: Liquid. In borate buffered saline, pH 8.4, containing 25mM sodium borate, 100mM boric acid, 75mM sodium chloride, 5mM EDTA (disodium salt). Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7708