Learn More
Biotium MemBrite™ Fix 405/430 Cell Surface Staining Kit, 100 Reactions
Catalog No. NC1535789
Encompass_Preferred
MemBrite Fix 405/430 is a blue fluorescent dye that covalently labels the surface of live cells. Unlike lipophilic membrane dyes, MemBrite is well-retained after fixation and permeabilization or methanol fixation. One vial for 100 labeling reactions (200 uL staining volume). Ex/Em 404/431 nm.
Catalog No. NC1535789 Supplier Biotium Supplier No. 30092T
MemBrite Fix 405/430 is a blue fluorescent dye that covalently labels the surface of live cells. Unlike lipophilic membrane dyes, MemBrite is well-retained after fixation and permeabilization or methanol fixation. One vial for 100 labeling reactions (200 uL staining volume). Ex/Em 404/431 nm.