Learn More
Biotium MemBrite™ Fix 488/515 Cell Surface Staining Kit, 500 Reactions
Catalog No. 501964512
Encompass_Preferred
MemBrite™ Fix Cell Surface Staining Kits are designed for covalently staining the surface of live cells to conveniently visualize cell boundaries in immunofluorescence experiments. MemBrite™ Fix are new covalent cell surface stains that react irreversibly with cell surface proteins for staining that can withstand formaldehyde or alcohol fixation, and detergent permeabilization. Unlike lectins such as WGA, which bind specific targets that may vary between cell types, MemBrite™ Fix dyes react widely with cell surface proteins. MemBrite™ Fix staining is rapid and non-toxic to cells, and because MemBrite™ Fix dyes are highly water soluble, they stain cells much more evenly than traditional lipophilic membrane dyes like DiO, DiI, PKH, Vybrant, or CellMask™. This kit has reagents sufficient for 500 labeling reactions. The kit contains 5 vials of MemBrite™ Fix 488/515. Each vial is sufficient for 20 uL of 1000X dye after DMSO reconstitution.
Catalog No. 50-196-4512 Supplier Biotium Supplier No. 30093
MemBrite™ Fix Cell Surface Staining Kits are designed for covalently staining the surface of live cells to conveniently visualize cell boundaries in immunofluorescence experiments. MemBrite™ Fix are new covalent cell surface stains that react irreversibly with cell surface proteins for staining that can withstand formaldehyde or alcohol fixation, and detergent permeabilization. Unlike lectins such as WGA, which bind specific targets that may vary between cell types, MemBrite™ Fix dyes react widely with cell surface proteins. MemBrite™ Fix staining is rapid and non-toxic to cells, and because MemBrite™ Fix dyes are highly water soluble, they stain cells much more evenly than traditional lipophilic membrane dyes like DiO, DiI, PKH, Vybrant, or CellMask™. This kit has reagents sufficient for 500 labeling reactions. The kit contains 5 vials of MemBrite™ Fix 488/515. Each vial is sufficient for 20 uL of 1000X dye after DMSO reconstitution.