Learn More
Biotium MemBrite™ Fix 594/615 Cell Surface Staining Kit, 100 Reactions
Catalog No. 501964515
Encompass_Preferred
MemBrite™ Fix Cell Surface Staining Kits are designed for covalently staining the surface of live cells to conveniently visualize cell boundaries in immunofluorescence experiments. MemBrite™ Fix are new covalent cell surface stains that react irreversibly with cell surface proteins for staining that can withstand formaldehyde or alcohol fixation, and detergent permeabilization. Unlike lectins such as WGA, which bind specific targets that may vary between cell types, MemBrite™ Fix dyes react widely with cell surface proteins. MemBrite™ Fix staining is rapid and non-toxic to cells, and because MemBrite™ Fix dyes are highly water soluble, they stain cells much more evenly than traditional lipophilic membrane dyes like DiO, DiI, PKH, Vybrant, or CellMask™. This trial size kit has reagents sufficient for 100 labeling reactions. The kit contains 1 vial of MemBrite™ Fix 594/615 that is sufficient for 20 uL of 1000X dye after DMSO reconstitution.
Catalog No. 50-196-4515 Supplier Biotium Supplier No. 30096T
MemBrite™ Fix Cell Surface Staining Kits are designed for covalently staining the surface of live cells to conveniently visualize cell boundaries in immunofluorescence experiments. MemBrite™ Fix are new covalent cell surface stains that react irreversibly with cell surface proteins for staining that can withstand formaldehyde or alcohol fixation, and detergent permeabilization. Unlike lectins such as WGA, which bind specific targets that may vary between cell types, MemBrite™ Fix dyes react widely with cell surface proteins. MemBrite™ Fix staining is rapid and non-toxic to cells, and because MemBrite™ Fix dyes are highly water soluble, they stain cells much more evenly than traditional lipophilic membrane dyes like DiO, DiI, PKH, Vybrant, or CellMask™. This trial size kit has reagents sufficient for 100 labeling reactions. The kit contains 1 vial of MemBrite™ Fix 594/615 that is sufficient for 20 uL of 1000X dye after DMSO reconstitution.