Learn More
Biotium MemBrite™ Fix-ST 650/665 Cell Surface Staining Kit, 100 Reactions
Supplier:  Biotium 30101T
Encompass_Preferred
MemBrite™ Fix Cell Surface Staining Kits are designed for covalently staining the surface of live cells to conveniently visualize cell boundaries in immunofluorescence experiments. MemBrite™ Fix are new covalent cell surface stains that react irreversibly with cell surface proteins for staining that can withstand formaldehyde or alcohol fixation, and detergent permeabilization. MemBrite™ Fix staining is rapid and non-toxic to cells, and because the dyes are highly water soluble, they stain cells much more evenly than traditional lipophilic membrane dyes like DiO, DiI, PKH, Vybrant, or CellMask™. MemBrite™ Fix-ST 650/665 is recommended for super-resolution imaging by STORM. This trial size kit has reagents sufficient for 100 labeling reactions. The kit contains 1 vial of MemBrite™ Fix-ST 650/665 that is sufficient for 20 uL of 1000X dye after DMSO reconstitution.
Catalog No. 50-196-4522