Learn More
EZKEM Memmbrane 25 mm Teflon Gas Diffusion 25 pk
SDP
Catalog No. NC0875493
Encompass_Preferred
EZkem’s 25mm Teflon Gas Diffusion Membranes are used on gas diffusion modules for chloroform extraction. (25 Pack)
Catalog No. NC0875493 Supplier EZKEM Supplier No. A002040
EZkem’s 25mm Teflon Gas Diffusion Membranes are used on gas diffusion modules for chloroform extraction. (25 Pack)