Learn More
MDI Membrane MEMBRN SCN 0.3MX3M 0.45UM ROLL
Supplier:  MDI Membrane SCNX8402XXXX101
Encompass_Preferred
mdi Cellulose Nitrate Blotting membrane, 300mm X 3m roll, 0.45 µm, non sterile, additional sizes available upon request, 1 ROLL
Catalog No. 50-109-8781