Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beckman Coulter™ MeMed BV™ Calibrator

Catalog No. 21402180 Shop All Beckman Coulter Products
Change view
Click to view available options
Quantity:
1.5 mL/Vial
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-402-180 1.5 mL/Vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-402-180 Supplier Beckman Coulter™ Supplier No. D15542
Add to Cart
Add to Cart

Calibrator material for the MeMed BV™ test on the MeMed Key™

  • Calibration material designed for use with the MeMed BV™ assay on Beckman Coulter systems to ensure accurate and reliable test performance
  • Specifically formulated calibrator for the MeMed BV™ assay, ensuring precise alignment and accuracy on Beckman Coulter analyzers
  • Supports consistent assay performance by maintaining reliable calibration across testing runs
  • Optimized for compatibility with Beckman Coulter systems, enabling seamless integration into laboratory workflows
  • Helps improve diagnostic confidence through stable and dependable calibration results
TRUSTED_SUSTAINABILITY

Specifications

Sample Type Whole Blood, Serum, Plasma
Type Clinical Chemistry Reagents
Disposable Single-use
Level 1, 2, 3
Quantity 1.5 mL/Vial
Sterility Sterile
Form Liquid Ready-to-Use

Sold in partnership with Beckman Coulter Diagnostics

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.