Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beckman Coulter™ MeMed BV™ External Control

Catalog No. 21402181 Shop All Beckman Coulter Products
Change view
Click to view available options
Quantity:
10 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-402-181 10 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-402-181 Supplier Beckman Coulter™ Supplier No. D15543
Add to Cart
Add to Cart

Quality control material for the MeMed BV™ test on the MeMed Key™

  • Designed as an external quality control material for the MeMed BV™ test, ensuring reliable verification of assay performance
  • Compatible with the MeMed Key™ platform for seamless and accurate quality control procedures
  • Supports consistent monitoring of test accuracy and system performance in laboratory settings
  • Helps laboratories maintain confidence in diagnostic results through dependable quality control processes
TRUSTED_SUSTAINABILITY

Specifications

Sample Type Whole Blood, Serum, Plasma
Type Clinical Chemistry Reagents
Disposable Single-use
Level 1, 2
Quantity 10 Tests
Form Liquid Ready-to-Use

Sold in partnership with Beckman Coulter Diagnostics

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.