Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beckman Coulter™ MeMed BV™ Test Cartridges, 96 DET

Catalog No. 21402179 Shop All Beckman Coulter Products
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-402-179 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-402-179 Supplier Beckman Coulter™ Supplier No. D15541
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Host immune response assay for accurately distinguishing between bacterial and viral infections in the ED workflow.

  • In just 15 minutes, direct from a small whole blood or serum sample, MeMed BV computationally integrates the levels of three host immune proteins into a simple score indicating the likelihood of a bacterial immune response or co-infection versus a likely viral immune response
  • Together with the MeMed KeyTM immunoassay instrument, MeMed BV helps physicians make better decisions in the management of patients with acute infections in a timely manner
  • Detects the body's immune response to a pathogen
  • Time to first result is faster than traditional methods
  • More accurate than CRP and Procalcitonin alone
  • Supports better antibiotic decision-making
  • Test can be utilized in both adult and pediatric populations
TRUSTED_SUSTAINABILITY

Specifications

CE Marker CE-IVD
Clia Complexity Moderate
DoA Calibrators No
Type Clinical Chemistry Reagents
Disposable Single-use
Quantity 96 Tests
Form Liquid Ready-to-Use

Sold in partnership with Beckman Coulter Diagnostics

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.