Learn More
Beckman Coulter™ MeMed Key™ Analyzer
Catalog No. 21402182 Shop All Beckman Coulter Products
Change view
Click to view available options
Type:
Automated Enzyme Immunoassay Analyzer
Catalog No. Type
21-402-182 Automated Enzyme Immunoassay Analyzer
1 options
Catalog No. 21-402-182 Supplier Beckman Coulter™ Supplier No. D15767
Add to Cart
Add to Cart
Compact, easy-to-use immunoassay platform that delivers rapid, highly accurate results to support critical clinical decisions.
  • Enables low volume sample types due to its miniaturized chemiluminescence detection technology, MeMed Key analyzes low volumes - pg/mL level detection
  • Low-Maintenance - does not require loading of chemical reagents, consumables, waste handling or other maintenance duties. Just insert the cartridge with the patient sample and wait for the result.
  • Easy to use Graphical User Interface (GUI), sample-to-result workflow and connects seamlessly with LIS systems.
  • Small Footprint 9.84 (H) x 9.84 (W) x 13.4 in. (D), 17 lb.
  • On-board Cartridge contains all that is required to conduct the test, as well as a safe waste compartment which frees the user from handling hazardous materials.
  • Designed to run the MeMed BV™ test in just 15 minutes from serum or venous whole blood to distinguish between bacterial or viral infections in both pediatric and adult populations.
TRUSTED_SUSTAINABILITY
Specifications
Type Automated Enzyme Immunoassay Analyzer
Sold in partnership with Beckman Coulter Diagnostics