Learn More
Tisch Scientific Tisch Scientific MCE Membrane Filters, Black w/White Grid, 0.45 µm, 25 mm, Nonsterile, 100 Pack, SF14731
SDP
Catalog No. NC2076512
Encompass
Tisch MCE membrane filters are developed from a mixture of cellulose acetate and cellulose nitrate and are one of the most widely used filters in both air monitoring and analytical applications. These filters are gridded for microbial cell counting and particle detection.
Catalog No. NC2076512 Supplier Tisch Scientific Supplier No. SF14731
Tisch MCE membrane filters are developed from a mixture of cellulose acetate and cellulose nitrate and are one of the most widely used filters in both air monitoring and analytical applications. These filters are gridded for microbial cell counting and particle detection.