Learn More
Cytoskeleton, Inc. MemGlow™ 488 Fluorogenic Plasma Membrane Probe, High Specificity, Non-Toxic, 2 nmol
Catalog No. NC1950107
Encompass
MemGlow™ 488 is a fluorogenic plasma membrane probe designed for high-specificity, low-background imaging of live and fixed cells. Its unique self-quenched "turn-on" mechanism allows fluorescence only upon membrane integration, offering clear visualization of cellular structures. Non-toxic and versatile, it supports various microscopy techniques, making it ideal for dynamic studies and long-term imaging applications.
Catalog No. NC1950107 Supplier Cytoskeleton, Inc. Supplier No. MG0102
MemGlow™ 488 is a fluorogenic plasma membrane probe designed for high-specificity, low-background imaging of live and fixed cells. Its unique self-quenched "turn-on" mechanism allows fluorescence only upon membrane integration, offering clear visualization of cellular structures. Non-toxic and versatile, it supports various microscopy techniques, making it ideal for dynamic studies and long-term imaging applications.