Learn More
Molecular Dimensions MEMGOLD 1/2 HT VALUE PACK
SDP
Catalog No. NC0426198
Encompass
Protein Crystallography. MEMGOLD screen COMBO PACK HT-96 (192 x 1.1mL): To give you the best start with crystallization of your membrane proteins we put together our two best selling membrane protein screens - MemGold and MemGold2 - in this special Combo Value Pack.
Catalog No. NC0426198 Supplier Molecular Dimensions Supplier No. MD174HT
Protein Crystallography. MEMGOLD screen COMBO PACK HT-96 (192 x 1.1mL): To give you the best start with crystallization of your membrane proteins we put together our two best selling membrane protein screens - MemGold and MemGold2 - in this special Combo Value Pack.