Learn More
Molecular Dimensions MEMGOLD 2 -- HT 96 CRYSTALLIZA
SDP
Catalog No. NC0351764
Encompass
Protein Crystallography. MemGold2 screen HT-96 (96 x 1.1mL): features 96 of the most recent alpha helical membrane protein crystallization conditions data mined from the PDB. For Prokaryotic and Eukaryotic alpha helical membrane proteins. Wide range of pH's (4 - 10). Small MW PEGs.
Catalog No. NC0351764 Supplier Molecular Dimensions Supplier No. MD164
Add to Cart
Add to Cart
Protein Crystallography. MemGold2 screen HT-96 (96 x 1.1mL): features 96 of the most recent alpha helical membrane protein crystallization conditions data mined from the PDB. For Prokaryotic and Eukaryotic alpha helical membrane proteins. Wide range of pH's (4 - 10). Small MW PEGs.