Learn More
Molecular Dimensions MemGoldMeso
SDP
Catalog No. NC1604550
Encompass
Developed from the most up-to-date survey of successful LCP crystallisation available: Analysis includes all mesophase structures in the protein database before March 2017. More structures were used for the identification of optimum in meso crystallisation conditions. Conditions were taken from a hand-curated database to ensure no false positives from unrelated structures contributed to the formulation. The recent analysis means a wider range of proteins were used in screen development, reducing bias to GPCR and rhodopsin conditions.
Catalog No. NC1604550 Supplier Molecular Dimensions Supplier No. MD1115
Developed from the most up-to-date survey of successful LCP crystallisation available: Analysis includes all mesophase structures in the protein database before March 2017. More structures were used for the identification of optimum in meso crystallisation conditions. Conditions were taken from a hand-curated database to ensure no false positives from unrelated structures contributed to the formulation. The recent analysis means a wider range of proteins were used in screen development, reducing bias to GPCR and rhodopsin conditions.