Learn More
Memmert Memmert Constant Climate Chamber HPP260Eco. 260L - 115V, Stainless Steel Construction with TwinDISPLAY digital controller
SDP
Catalog No. 502254405
Encompass_Preferred
Memmert Constant Climate Chamber (HPPeco) - this programmable model is the 115 volt, Peltier-technology seamlessly heats and cools, with TwinDISPLAY digital controller. Includes (2) Stainless Steel Grid Shelves, Atmocontrol Software, Atmocontrol Manual, Operation manual, Calibration Certificate and Power Cord - energy efficient, precise. Setting temperature range +5C to +70C. Setting humidity range 10%RH - 90%RH. Internal dimensions 640/800/468-W/H/D mm.
Catalog No. 50-225-4405 Supplier Memmert Supplier No. HPP260ECO 115V
Add to Cart
Add to Cart
Request a Quote
Memmert Constant Climate Chamber (HPPeco) - this programmable model is the 115 volt, Peltier-technology seamlessly heats and cools, with TwinDISPLAY digital controller. Includes (2) Stainless Steel Grid Shelves, Atmocontrol Software, Atmocontrol Manual, Operation manual, Calibration Certificate and Power Cord - energy efficient, precise. Setting temperature range +5C to +70C. Setting humidity range 10%RH - 90%RH. Internal dimensions 640/800/468-W/H/D mm.