Learn More
Memmert Memmert Stability Climate Chamber ICH110Eco. 110L - 115V, Stainless Steel Construction with TwinDISPLAY Controller
SDP
Catalog No. 502254401
Encompass_Preferred
Memmert Climate Chamber - this programmable model is the 115 volt, for stability testing according to ICH guidelines, with TwinDISPLAY digital controller. ICH humidity range is 10 to 80% rh (Optional Light or CO2 modules available with customization). Each individual appliance is tested according to the requirements of DIN 12 880: 2007-05. Includes (2) Stainless Steel Grid Shelves, Atmocontrol Software, Atmocontrol Manual, Operation manual, Calibration Certificate and Power Cord - unparalleled stability!
This item is not returnable. View return policy
Catalog No. 50-225-4401 Supplier Memmert Supplier No. ICH110 115V
Add to Cart
Add to Cart
Request a Quote
Memmert Climate Chamber - this programmable model is the 115 volt, for stability testing according to ICH guidelines, with TwinDISPLAY digital controller. ICH humidity range is 10 to 80% rh (Optional Light or CO2 modules available with customization). Each individual appliance is tested according to the requirements of DIN 12 880: 2007-05. Includes (2) Stainless Steel Grid Shelves, Atmocontrol Software, Atmocontrol Manual, Operation manual, Calibration Certificate and Power Cord - unparalleled stability!