Learn More
Memmert Memmert Incubator IF55. 55L - 115V, Forced-Air Convection, Stainless Steel Construction with SingleDISPLAY digital controller.
SDP
Catalog No. 502254345
Encompass_Preferred
Memmert Incubators - this model is the 115 volt, forced-air convection, with SingleDISPLAY digital controller. Interior dimensions 400/400/291-W/H/D mm. Includes (1) Stainless Steel Grid Shelf, Quick Start Guide, Calibration Certificate and Power Cord - Working Range 10C above ambient up to +80C.
This item is not returnable. View return policy
Catalog No. 50-225-4345 Supplier Memmert Supplier No. IF55
Memmert Incubators - this model is the 115 volt, forced-air convection, with SingleDISPLAY digital controller. Interior dimensions 400/400/291-W/H/D mm. Includes (1) Stainless Steel Grid Shelf, Quick Start Guide, Calibration Certificate and Power Cord - Working Range 10C above ambient up to +80C.