Learn More
Memmert Memmert Incubator IF260Plus. 260L - 115V, Forced-Air Convection, Stainless Steel Construction with TwinDISPLAY digital controller.
SDP
Catalog No. 502254363
Encompass_Preferred
Memmert Incubators - this programmable model is the 115 volt, forced-air convection, with TwinDISPLAY digital controller. Interior dimensions 640/800/461-W/H/D mm. Includes (2) Stainless Steel Grid Shelves, Atmocontrol Software, Manual, Quick Start Guide, Calibration Certificate and Power Cord - Working Range 10C above ambient to +300C.
This item is not returnable. View return policy
Catalog No. 50-225-4363 Supplier Memmert Supplier No. IF260 PLUS
Add to Cart
Add to Cart
Request a Quote
Memmert Incubators - this programmable model is the 115 volt, forced-air convection, with TwinDISPLAY digital controller. Interior dimensions 640/800/461-W/H/D mm. Includes (2) Stainless Steel Grid Shelves, Atmocontrol Software, Manual, Quick Start Guide, Calibration Certificate and Power Cord - Working Range 10C above ambient to +300C.