Learn More
Memmert Memmert Humidity Chamber HCP50 - 50L - 115V, Stainless Steel Construction with Programmable TwinDISPLAY controller
SDP
Catalog No. 502254397
Encompass_Preferred
Memmert Humidity Chamber - HCP provides excellent control, making it easy to achieve uniform atmosphere and temperature distribution in simulating environmental condistions. This 56L (2 cu ft) model is the 115 volt, with working humidity ranges of 20 to 95% rh, within temperatures of at least 6¿C over ambient up to 90¿C with humidity control. Includes (1) Stainless Steel Perforated Shelf, Atmocontrol Software + Manual, Operation manual, Calibration Certificate, Power Cord, Water tank and connections!
This item is not returnable. View return policy
Catalog No. 50-225-4397 Supplier Memmert Supplier No. HCP 50 115V
Add to Cart
Add to Cart
Request a Quote
Memmert Humidity Chamber - HCP provides excellent control, making it easy to achieve uniform atmosphere and temperature distribution in simulating environmental condistions. This 56L (2 cu ft) model is the 115 volt, with working humidity ranges of 20 to 95% rh, within temperatures of at least 6¿C over ambient up to 90¿C with humidity control. Includes (1) Stainless Steel Perforated Shelf, Atmocontrol Software + Manual, Operation manual, Calibration Certificate, Power Cord, Water tank and connections!