Learn More
Labrepco Inc Memmert HCP150 Humidity Chamber, 156 L Capacity, Temperature Range +18 °C to +90 °C, 3-Year Warranty
SDP
Catalog No. NC3652056
Encompass
The Memmert HCP150 Humidity Chamber is designed for precise environmental testing, enabling rapid temperature and humidity adjustments for optimal sample integrity. Its durable stainless steel construction, advanced control system, and comprehensive safety features make it suitable for various scientific applications. Ideal for biology, electronics, and building physics, this chamber stands out with its efficient heating system and user-friendly interface, ensuring reliable performance in demanding conditions.
Catalog No. NC3652056 Supplier Labrepco Inc Supplier No. HCP150
Add to Cart
Add to Cart
Request a Quote
The Memmert HCP150 Humidity Chamber is designed for precise environmental testing, enabling rapid temperature and humidity adjustments for optimal sample integrity. Its durable stainless steel construction, advanced control system, and comprehensive safety features make it suitable for various scientific applications. Ideal for biology, electronics, and building physics, this chamber stands out with its efficient heating system and user-friendly interface, ensuring reliable performance in demanding conditions.