Learn More
Memmert Memmert Universal Heating and Drying Laboratory Oven UN30. 30L-115 Volt, Natural Convection, Stainless Steel Construction with SingleDISPLAY Digital Controller.
SDP
Catalog No. 502254245
Encompass_Preferred
Memmert Universal heating and drying Oven - this model is the 115 volt, natural convection, with SingleDISPLAY digital controller. Interior dimensions 400/320/250-W/H/D mm. Includes (1) Stainless Steel Grid Shelf, Quick Start Guide, Calibration Certificate and Power Cord - Working Range 5C above ambient to +300C.
This item is not returnable. View return policy
Catalog No. 50-225-4245 Supplier Memmert Supplier No. UN30
Memmert Universal heating and drying Oven - this model is the 115 volt, natural convection, with SingleDISPLAY digital controller. Interior dimensions 400/320/250-W/H/D mm. Includes (1) Stainless Steel Grid Shelf, Quick Start Guide, Calibration Certificate and Power Cord - Working Range 5C above ambient to +300C.