Learn More
Memmert Memmert Incubator IN55. 55L - 230V, Natural Convection, Stainless Steel Construction with SingleDISPLAY digital controller.
SDP
Catalog No. 502254314
Encompass_Preferred
Memmert Incubators - this model is the 230 volt, natural convection, with SingleDISPLAY digital controller. Interior dimensions 400/400/330-W/H/D mm. Includes (1) Stainless Steel Grid Shelf, Quick Start Guide, Calibration Certificate and Power Cord - Working Range 5C above ambient up to +80C.
This item is not returnable. View return policy
Catalog No. 50-225-4314 Supplier Memmert Supplier No. IN55 230V
Memmert Incubators - this model is the 230 volt, natural convection, with SingleDISPLAY digital controller. Interior dimensions 400/400/330-W/H/D mm. Includes (1) Stainless Steel Grid Shelf, Quick Start Guide, Calibration Certificate and Power Cord - Working Range 5C above ambient up to +80C.