Learn More
Memmert Memmert Incubator IN450Plus. 450L - 115V, Natural Convection, Stainless Steel Construction with TwinDISPLAY digital controller.
SDP
Catalog No. 502254335
Encompass_Preferred
Memmert Incubators - this programmable model is the 115 volt, natural convection, with TwinDISPLAY digital controller. Interior dimensions 1040/720/600-W/H/D mm. Includes (2) Stainless Steel Grid Shelves, Quick Start Guide, Atmocontrol Software, Manual, Calibration Certificate and Power Cord - Working Range 5C above ambient up to +80C.
This item is not returnable. View return policy
Catalog No. 50-225-4335 Supplier Memmert Supplier No. IN450 PLUS
Add to Cart
Add to Cart
Request a Quote
Memmert Incubators - this programmable model is the 115 volt, natural convection, with TwinDISPLAY digital controller. Interior dimensions 1040/720/600-W/H/D mm. Includes (2) Stainless Steel Grid Shelves, Quick Start Guide, Atmocontrol Software, Manual, Calibration Certificate and Power Cord - Working Range 5C above ambient up to +80C.