Learn More
Memmert Memmert Incubator IN75. 75L - 230V, Natural Convection, Stainless Steel Construction with SingleDISPLAY digital controller.
SDP
Supplier:  Memmert IN75 230V
Encompass_Preferred
Memmert Incubators - this model is the 230 volt, natural convection, with SingleDISPLAY digital controller. Interior dimensions 400/560/330-W/H/D mm. Includes (2) Stainless Steel Grid Shelves, Quick Start Guide, Calibration Certificate and Power Cord - Working Range 5C above ambient up to +80C.
Catalog No. 50-225-4318