Learn More
Memmert Memmert Incubator IN750. 750L - 230V, Natural Convection, Stainless Steel Construction with SingleDISPLAY digital controller.
SDP
Catalog No. 502254338
Encompass_Preferred
Memmert Incubators - this model is the 230 volt, natural convection, with SingleDISPLAY digital controller. Interior dimensions 1040/1200/600-W/H/D mm. Includes (2) Stainless Steel Grid Shelves, Quick Start Guide, Calibration Certificate and Power Cord - Working Range 5C above ambient up to +80C.
This item is not returnable. View return policy
Catalog No. 50-225-4338 Supplier Memmert Supplier No. IN750 230V
Add to Cart
Add to Cart
Request a Quote
Memmert Incubators - this model is the 230 volt, natural convection, with SingleDISPLAY digital controller. Interior dimensions 1040/1200/600-W/H/D mm. Includes (2) Stainless Steel Grid Shelves, Quick Start Guide, Calibration Certificate and Power Cord - Working Range 5C above ambient up to +80C.