Learn More
Memmert MEMMERT V029-230VOLT VACM OVEN
SDP
Catalog No. NC3441853
Encompass_Preferred
  • High-performance vacuum oven for drying sensitive materials.
  • Precise temperature and digital pressure control for reproducible results.
  • Multi-level heating and sensing system provides uniform temperature distribution.
  • Individually controlled thermoshelves minimize heat loss.
  • TwinDISPLAY control panel with dual high-definition TFT displays.
  • Fully programmable via AtmoCONTROL software for process management.
  • Durable, corrosion-resistant stainless steel construction.
  • Optional speed-controlled vacuum pump enhances energy efficiency by 70%.
  • Chamber volume of 29 L (1.0 cu. ft.).
  • Temperature range from ambient +5°C to 200°C.
  • Vacuum range of 5 to 1100 mbar, with permitted final vacuum of 0.01 mbar.
  • Digital timer from 1 minute to 99 days.
  • Safety features include thermal monitoring and alarms.
  • 3-year warranty for reliability.
Catalog No. NC3441853 Supplier Memmert Supplier No. VO 29 230VOLT
Add to Cart
Add to Cart
Request a Quote
  • High-performance vacuum oven for drying sensitive materials.
  • Precise temperature and digital pressure control for reproducible results.
  • Multi-level heating and sensing system provides uniform temperature distribution.
  • Individually controlled thermoshelves minimize heat loss.
  • TwinDISPLAY control panel with dual high-definition TFT displays.
  • Fully programmable via AtmoCONTROL software for process management.
  • Durable, corrosion-resistant stainless steel construction.
  • Optional speed-controlled vacuum pump enhances energy efficiency by 70%.
  • Chamber volume of 29 L (1.0 cu. ft.).
  • Temperature range from ambient +5°C to 200°C.
  • Vacuum range of 5 to 1100 mbar, with permitted final vacuum of 0.01 mbar.
  • Digital timer from 1 minute to 99 days.
  • Safety features include thermal monitoring and alarms.
  • 3-year warranty for reliability.