Learn More
Molecular Dimensions MEMMESO HT-96 (1.1 ML)
SDP
Catalog No. NC0660166
Encompass_Preferred
Protein Crystallography. MEMMESO screen HT-96 (96 x 1.1mL): Optimized to work in synergy with Lipidic Cubic Phase (LCP) and the LCP crystallization method, covering a range of pH, precipitants and salt. Up-to-date conditions data-mined from current reported GPCR crystal structures.
Catalog No. NC0660166 Supplier Molecular Dimensions Supplier No. MD187
Add to Cart
Add to Cart
Protein Crystallography. MEMMESO screen HT-96 (96 x 1.1mL): Optimized to work in synergy with Lipidic Cubic Phase (LCP) and the LCP crystallization method, covering a range of pH, precipitants and salt. Up-to-date conditions data-mined from current reported GPCR crystal structures.