Learn More
Cayman Chemical Memantine Lactose Adduct, Purity: ≥95%, CAS: 1159637-28-1, 5 mg
SDP
Catalog No. 502984229
Encompass_Preferred
A lactose adduct of memantine; intermediate in the synthesis of adamantane amine antibiotics
Catalog No. 50-298-4229 Supplier Cayman Chemical Supplier No. 206625 MG
A lactose adduct of memantine; intermediate in the synthesis of adamantane amine antibiotics