Learn More
Aviva Systems Biology Corp MEMO1 Antibody - middle region (ARP56786_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56786_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MEMO1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MEMO1. Alternate names: C2orf4, CGI-27, DKFZp434I0135, FLJ25031, MEMO, NS5ATP7ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61988